AMBH3155FC6B Ambeed, Inc.

4'-(4-Bromophenyl)-2,2':6',2''-terpyridine

  • MDL number MFCD06796984
  • Cas Number : 89972-76-9
  • Molecular Weight : 388.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 162�t N/A mmHg
bp 512.671�t 760 mmHg
Iupac Name 4'-(4-bromophenyl)-2,2':6',2''-terpyridine
Inchi Code 1S/C21H14BrN3/c22-17-9-7-15(8-10-17)16-13-20(18-5-1-3-11-23-18)25-21(14-16)19-6-2-4-12-24-19/h1-14H
Inchi Key BOPPHKMZOYPASP-UHFFFAOYSA-N

Description

Melting Point 162�t N/A mmHg
Purity 97%
Flash Point 263.854
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 512.671�t 760 mmHg
Product Link https://www.ambeed.com/products/89972-76-9.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/89972-76-9.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart