AMBH3155FCA2 Ambeed, Inc.

N-(2-Methoxyphenyl)-3-oxobutanamide

  • MDL number MFCD00008781
  • Cas Number : 92-15-9
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-87�t N/A mmHg
Purity 98
Flash Point 179.3
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 372.9�t 760 mmHg
Product Link https://www.ambeed.com/products/92-15-9.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312+P330-P305+P351+P338+P337+P313-P501
MSDS https://www.ambeed.com/products/92-15-9.html
pictograms GHS07
hstatements H302-H320
Availability details for
Quantity
  • Availability
  • Add to Cart