AMBH3155FCA3 Ambeed, Inc.

N-(5-Fluoro-2-oxo-2,3-dihydropyrimidin-4-yl)benzamide

  • MDL number MFCD27941084
  • Cas Number : 10357-07-0
  • Molecular Weight : 233.2000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/10357-07-0.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/10357-07-0.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart