AMBH3155FCB5 Ambeed, Inc.

tert-Butyl 4-amino-2-oxo-5,6-dihydropyridine-1(2H)-carboxylate

  • MDL number MFCD18207247
  • Cas Number : 1333319-62-2
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-amino-6-oxo-3,6-dihydro-1(2H)-pyridinecarboxylate
Inchi Code 1S/C10H16N2O3/c1-10(2,3)15-9(14)12-5-4-7(11)6-8(12)13/h6H,4-5,11H2,1-3H3
Inchi Key RNSZCMUVIYHTIU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1333319-62-2.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1333319-62-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart