AMBH3155FCCC Ambeed, Inc.

Potassium oleate

  • MDL number MFCD00064243
  • Cas Number : 143-18-0
  • Molecular Weight : 320.5500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/143-18-0.html

Safety Information

signalword Danger
pcodes P261-P273-P305+P351+P338
MSDS https://www.ambeed.com/products/143-18-0.html
pictograms GHS07,GHS09
hstatements H315-H319-H335-H400
Availability details for
Quantity
  • Availability
  • Add to Cart