AMBH3155FCD8 Ambeed, Inc.

Sodium Dimethyldithiocarbamate Dihydrate

  • MDL number MFCD00150729
  • Cas Number : 72140-17-1
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 120-122�t N/A mmHg
Iupac Name sodium dimethyldithiocarbamate dihydrate
Inchi Code 1S/C3H7NS2.Na.2H2O/c1-4(2)3(5)6;;;/h1-2H3,(H,5,6);;2*1H2/q;+1;;/p-1
Inchi Key NQVBYQPOGVWUPK-UHFFFAOYSA-M

Description

Melting Point 120-122�t N/A mmHg
Purity 98%
Flash Point 32
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/72140-17-1.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/72140-17-1.html
pictograms GHS09
hstatements H400
Availability details for
Quantity
  • Availability
  • Add to Cart