AMBH3155FCF0 Ambeed, Inc.

Methyl 2-amino-5-chloro-4-methoxybenzoate

  • MDL number MFCD24626483
  • Cas Number : 79025-26-6
  • Molecular Weight : 215.6300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79025-26-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/79025-26-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart