AMBH3155FCF8 Ambeed, Inc.

2-(3-Bromophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036584
  • Cas Number : 90433-20-8
  • Molecular Weight : 224.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-bromophenyl)-2-methylpropanenitrile
Inchi Code 1S/C10H10BrN/c1-10(2,7-12)8-4-3-5-9(11)6-8/h3-6H,1-2H3
Inchi Key NKECRCMIFURWMK-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/90433-20-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/90433-20-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart