AMBH3155FCFC Ambeed, Inc.

3,4-Diaminothiophene

  • MDL number MFCD00066983
  • Cas Number : 78637-85-1
  • Molecular Weight : 114.1700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/78637-85-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/78637-85-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart