AMBH3155FD07 Ambeed, Inc.

2-Bromo-1,5-difluoro-3-methylbenzene

  • MDL number MFCD28738349
  • Cas Number : 1807135-08-5
  • Molecular Weight : 207.0200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1807135-08-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1807135-08-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart