AMBH3155FD11 Ambeed, Inc.

2-Amino-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD28359825
  • Cas Number : 1288999-05-2
  • Molecular Weight : 189.1346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1288999-05-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1288999-05-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart