AMBH3155FD19 Ambeed, Inc.

5-(Chloromethyl)nicotinonitrile

  • MDL number MFCD10697604
  • Cas Number : 562074-59-3
  • Molecular Weight : 152.5810

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/562074-59-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/562074-59-3.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart