AMBH3155FD24 Ambeed, Inc.

N-(5-Bromo-4-ethylthiazol-2-yl)acetamide

  • MDL number MFCD12756845
  • Cas Number : 1267904-60-8
  • Molecular Weight : 249.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(5-bromo-4-ethyl-1,3-thiazol-2-yl)acetamide
Inchi Code 1S/C7H9BrN2OS/c1-3-5-6(8)12-7(10-5)9-4(2)11/h3H2,1-2H3,(H,9,10,11)
Inchi Key LODCKIISMKDQFH-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1267904-60-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1267904-60-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart