AMBH3155FD39 Ambeed, Inc.

Phenyl(pyridin-4-yl)methanamine

  • MDL number MFCD06589768
  • Cas Number : 58088-57-6
  • Molecular Weight : 184.2400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58088-57-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/58088-57-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart