AMBH3155FD3E Ambeed, Inc.

3-Methoxy-4-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD16606204
  • Cas Number : 1227581-88-5
  • Molecular Weight : 192.1400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1227581-88-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1227581-88-5.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart