AMBH3155FD50 Ambeed, Inc.

Guanosine-5-triphosphoric aicd disodium salt

  • MDL number MFCD00083629
  • Cas Number : 56001-37-7
  • Molecular Weight : 567.1400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 89
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/56001-37-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338-P302+P352-P321-P405-P501
MSDS https://www.ambeed.com/products/56001-37-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart