AMBH3155FD6B Ambeed, Inc.

M-PEg5-bromide

  • MDL number MFCD28950766
  • Cas Number : 854601-80-2
  • Molecular Weight : 315.2000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form White to yellow powder or crystals or liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/854601-80-2.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/854601-80-2.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart