AMBH3155FD6F Ambeed, Inc.

N-Methyl-D-glucamine Hydrochloride [for Buffer]

  • MDL number MFCD00060164
  • Cas Number : 35564-86-4
  • Molecular Weight : 231.6800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/35564-86-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/35564-86-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart