AMBH3155FDA8 Ambeed, Inc.

5-Hydroxy-2-(hydroxymethyl)-1-phenylpyridin-4(1H)-one

  • MDL number MFCD27991853
  • Cas Number : 70033-61-3
  • Molecular Weight : 217.2200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/70033-61-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/70033-61-3.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart