AMBH3155FDB5 Ambeed, Inc.

Methyl 3-oxopyrrolidine-1-carboxylate

  • MDL number MFCD12195869
  • Cas Number : 69079-09-0
  • Molecular Weight : 143.1400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form Solid or liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69079-09-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69079-09-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart