AMBH3155FDCB Ambeed, Inc.

2,3-Dichloro-4-nitroaniline

  • MDL number MFCD11226295
  • Cas Number : 69951-03-7
  • Molecular Weight : 207.0100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/69951-03-7.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/69951-03-7.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart