AMBH3155FDCD Ambeed, Inc.

1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide

  • MDL number MFCD03788927
  • Cas Number : 174899-82-2
  • Molecular Weight : 391.3100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/174899-82-2.html

Safety Information

signalword Danger
pcodes P260-P264-P270-P273-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310-P305+P351+P338+P310-P405-P501
MSDS https://www.ambeed.com/products/174899-82-2.html
pictograms GHS05,GHS06
hstatements H301+H311-H314-H401
Availability details for
Quantity
  • Availability
  • Add to Cart