AMBH3155FDD6 Ambeed, Inc.

tert-Butyl 7-amino-2-azaspiro[4.5]decane-2-carboxylate

  • MDL number MFCD22033489
  • Cas Number : 1363382-19-7
  • Molecular Weight : 254.3700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form White to yellow powder or crystals liquid or semi-solid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1363382-19-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1363382-19-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart