AMBH3155FDD8 Ambeed, Inc.

Methyl 4-bromonaphthalene-1-carboxylate

  • MDL number MFCD09475914
  • Cas Number : 35615-97-5
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 370.765�t 760 mmHg
Iupac Name methyl 4-bromo-1-naphthoate
Inchi Code 1S/C12H9BrO2/c1-15-12(14)10-6-7-11(13)9-5-3-2-4-8(9)10/h2-7H,1H3
Inchi Key NSWYFJWAOXYZDF-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 178.033
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 370.765�t 760 mmHg
Product Link https://www.ambeed.com/products/35615-97-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/35615-97-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart