AMBH3155FDE2 Ambeed, Inc.

4-Oxo-3,4-dihydrothieno[2,3-d]pyrimidine-5-carbonitrile

  • MDL number MFCD30189012
  • Cas Number : 1801143-20-3
  • Molecular Weight : 177.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxythieno[2,3-d]pyrimidine-5-carbonitrile
Inchi Code 1S/C7H3N3OS/c8-1-4-2-12-7-5(4)6(11)9-3-10-7/h2-3H,(H,9,10,11)
Inchi Key KCLHVWPKZOEVON-UHFFFAOYSA-N

Description

Synonym 4-Hydroxythieno[2,3-d]pyrimidine-5-carbonitrile
Purity 97%
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1801143-20-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1801143-20-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart