AMBH3155FE07 Ambeed, Inc.

4,6-Dichloro-5-iodo-2-methylpyrimidine

  • MDL number MFCD21743354
  • Cas Number : 111079-21-1
  • Molecular Weight : 288.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,6-dichloro-5-iodo-2-methylpyrimidine
Inchi Code 1S/C5H3Cl2IN2/c1-2-9-4(6)3(8)5(7)10-2/h1H3
Inchi Key ISBJSJIFFXXZQH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/111079-21-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/111079-21-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart