AMBH3155FE09 Ambeed, Inc.

2-(2-Bromoethyl)-5-nitropyridine

  • MDL number N/A
  • Cas Number : 2229221-49-0
  • Molecular Weight : 231.0500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2229221-49-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/2229221-49-0.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart