AMBH3155FE10 Ambeed, Inc.

1-Chloro-3,5-di(4-chlorbenzoyl)-2-deoxy-D-ribose

  • MDL number MFCD00672145
  • Cas Number : 3601-90-9
  • Molecular Weight : 429.6784

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3601-90-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3601-90-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart