AMBH3155FE35 Ambeed, Inc.

1,3-Bis(aminopropyl)tetramethyldisiloxane

  • MDL number MFCD00039786
  • Cas Number : 2469-55-8
  • Molecular Weight : 248.5100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 83
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2469-55-8.html

Safety Information

signalword Danger
pcodes P210-P260-P264-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310-P305+P351+P338+P310-P370+P378-P403+P235-P405-P501
MSDS https://www.ambeed.com/products/2469-55-8.html
pictograms GHS05
hstatements H227-H314
Availability details for
Quantity
  • Availability
  • Add to Cart