AMBH3155FE45 Ambeed, Inc.

4-(3-(4-Hydroxyphenyl)-4,4-dimethyl-5-oxo-2-thioxoimidazolidin-1-yl)-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD28978634
  • Cas Number : 915086-32-7
  • Molecular Weight : 405.4000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/915086-32-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/915086-32-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart