AMBH324A4833 Ambeed, Inc.

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1H-imidazol-4-yl)propanoic acid

  • MDL number MFCD00190885
  • Cas Number : 116611-64-4
  • Molecular Weight : 377.3900

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/116611-64-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/116611-64-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart