AMBH324A48A8 Ambeed, Inc.

SMI-4a

  • MDL number MFCD01152003
  • Cas Number : 438190-29-5
  • Molecular Weight : 273.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/438190-29-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/438190-29-5.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart