AMBH324A4C86 Ambeed, Inc.

10058-F4

  • MDL number MFCD04969046
  • Cas Number : 403811-55-2
  • Molecular Weight : 249.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/403811-55-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/403811-55-2.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart