AMBH324A4D1D Ambeed, Inc.

8-(n-(4-methyl-1-piperazinyl)formidoyl)-rifomycins

  • MDL number MFCD00151389
  • Cas Number : 13292-46-1
  • Molecular Weight : 822.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/13292-46-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13292-46-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart