AMBH324A4D2B Ambeed, Inc.

(2R,3R)-3-((R)-1-((tert-Butyldimethylsilyl)oxy)ethyl)-4-oxoazetidin-2-yl acetate

  • MDL number MFCD00135115
  • Cas Number : 76855-69-1
  • Molecular Weight : 287.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/76855-69-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/76855-69-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart