AMBH324A4D30 Ambeed, Inc.

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((allyloxy)carbonyl)amino)propanoic acid

  • MDL number MFCD01076137
  • Cas Number : 178924-05-5
  • Molecular Weight : 410.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/178924-05-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/178924-05-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart