AMBH324A4D55 Ambeed, Inc.

(S)-tert-Butyl 3-amino-2-((tert-butoxycarbonyl)amino)propanoate

  • MDL number MFCD09263390
  • Cas Number : 77215-54-4
  • Molecular Weight : 260.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/77215-54-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/77215-54-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart