AMBH324A4D56 Ambeed, Inc.

Boc-Ala-D-Glu(OBzl)-NH2

  • MDL number MFCD30502864
  • Cas Number : 18814-49-8
  • Molecular Weight : 407.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/18814-49-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/18814-49-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart