AMBH324A4D61 Ambeed, Inc.

Z-D-Asp-OMe

  • MDL number MFCD09842077
  • Cas Number : 47087-37-6
  • Molecular Weight : 281.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/47087-37-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/47087-37-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart