AMBH324A4D64 Ambeed, Inc.

Tri-O-benzoyl-D-glucal

  • MDL number MFCD00061643
  • Cas Number : 13322-90-2
  • Molecular Weight : 458.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13322-90-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13322-90-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart