AMBH324A4D65 Ambeed, Inc.

Fmoc-D-3-Pal-OH

  • MDL number MFCD00080267
  • Cas Number : 142994-45-4
  • Molecular Weight : 388.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/142994-45-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/142994-45-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart