AMBH324A4D66 Ambeed, Inc.

(S)-4-Chloro-3-hydroxybutyronitrile

  • MDL number MFCD00273363
  • Cas Number : 127913-44-4
  • Molecular Weight : 119.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/127913-44-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/127913-44-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart