AMBH324A4D6B Ambeed, Inc.

tert-Butyl pitavastatin

  • MDL number MFCD12755990
  • Cas Number : 586966-54-3
  • Molecular Weight : 477.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/586966-54-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/586966-54-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart