AMBH324A4D75 Ambeed, Inc.

(S)-1-((S)-2-((Methoxycarbonyl)amino)-3-methylbutanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD27946414
  • Cas Number : 181827-47-4
  • Molecular Weight : 272.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/181827-47-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/181827-47-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart