AMBH324A4D76 Ambeed, Inc.

(R)-2-Amino-3-(benzyloxy)propan-1-ol hydrochloride

  • MDL number MFCD04112476
  • Cas Number : 58577-95-0
  • Molecular Weight : 217.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58577-95-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/58577-95-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart