AMBH324A4D77 Ambeed, Inc.

N1,N6-Bis(3-phenylallylidene)hexane-1,6-diamine

  • MDL number MFCD00046784
  • Cas Number : 140-73-8
  • Molecular Weight : 344.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/140-73-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/140-73-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart