AMBH324A4D82 Ambeed, Inc.

Fmoc-(S)-3-amino-3-(3-nitrophenyl)propanoic acid

  • MDL number MFCD01631072
  • Cas Number : 374791-01-2
  • Molecular Weight : 432.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/374791-01-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/374791-01-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart