AMBH324A4D93 Ambeed, Inc.

H-D-Asp(OBzl)-OH

  • MDL number MFCD00077100
  • Cas Number : 13188-89-1
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13188-89-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13188-89-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart