AMBH324A4D97 Ambeed, Inc.

D-Fructose 1,6-bisphosphate trisodium salt hydrate

  • MDL number MFCD00068321
  • Cas Number : 38099-82-0
  • Molecular Weight : 428.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/38099-82-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/38099-82-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart