AMBH324A4D9D Ambeed, Inc.

(S)-2-Amino-4-methylpentanamide

  • MDL number MFCD00238027
  • Cas Number : 687-51-4
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/687-51-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/687-51-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart